Merck
  • NIR-VCD, vibrational circular dichroism in the near-infrared: experiments, theory and calculations.
NIR-VCD, vibrational circular dichroism in the near-infrared: experiments, theory and calculations.
Chirality (2009-11-21)
Sergio Abbate, Ettore Castiglioni, Fabrizio Gangemi, Roberto Gangemi, Giovanna Longhi
ABSTRACT
The first well documented experiments of Near Infrared Vibrational Circular Dichroism (NIR-VCD) were performed around 1975. We review the thirty year history of NIR-VCD, encompassing both instrumental development and theoretical/computational methods that allow interpretation of experimental spectra, harvesting useful structural information therefrom. We hope to stimulate interest in this still scarcely explored spectroscopy of chiral molecules.
Sigma-Aldrich
Fenchyl alcohol, ≥96%, FG