Merck
  • Delta-di-carboxybutyl phosphoramidate of 2'-deoxycytidine-5'-monophosphate as substrate for DNA polymerization by HIV-1 reverse transcriptase.
Delta-di-carboxybutyl phosphoramidate of 2'-deoxycytidine-5'-monophosphate as substrate for DNA polymerization by HIV-1 reverse transcriptase.
Bioorganic & medicinal chemistry (2009-09-01)
Ivan Zlatev, Anne Giraut, François Morvan, Piet Herdewijn, Jean-Jacques Vasseur
ABSTRACT
The replacement of the pyrophosphate moiety of 2'-deoxynucleoside triphosphates by non natural delta-dicarboxylic butyl amino acid allows incorporation of natural 2'-deoxycytidine into DNA using HIV-1 reverse transcriptase (RT) as enzyme. In contrast, the 3'-deoxycytidine analogue was not a substrate of the HIV-1 RT.
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%