Merck
  • Possible involvement of 1,2-diacetylbenzene in diethylbenzene-induced neuropathy in rats.
Possible involvement of 1,2-diacetylbenzene in diethylbenzene-induced neuropathy in rats.
Journal of applied toxicology : JAT (1991-08-01)
F Gagnaire, A Ensminger, B Marignac, J De Ceaurriz
ABSTRACT
The role of 1,2-diacetylbenzene (1,2-DAB) in the peripheral nerve toxicity of 1,2-diethylbenzene (1,2-DEB) was investigated in rats. Gas chromatography-mass spectrometry identified 1,2-DAB in the urine samples of rats given 165 mg kg-1 1,2-DEB orally on four consecutive days. 1,2-DAB shared not only the ability of 1,2-DEB to cause bluish discoloration of skin, internal organs and urine, but unlike 1,2-DEB it turned hair blue at the site of intraperitoneal injection. Intraperitoneal administration of 10 mg kg-1 and 20 mg kg-1 1,2-DAB to groups of 12 rats, 4 days a week for 11 and 6 weeks, caused a dose- and time-dependent decrease in mean sensory and motor conduction velocities. Recovery in a 5-week post-exposure period was gradual but consistent. The effect of 1,2-DAB on the amplitude of the sensory action potential was ambiguous. The findings support the hypothesis that the formation of 1,2-diacetylbenzene derivatives contributes to the neurotoxicity of 1,2-DEB.
Sigma-Aldrich
1,2-Diacetylbenzene, 99%
Sigma-Aldrich
1,2-Diethylbenzene, technical grade, 92%
Sigma-Aldrich
1,2-Diethylbenzene, ≥99.0% (GC)