Merck
  • Stereoselective one-pot, three-component synthesis of 4-aryltetrahydropyran via Prins-Friedel-Crafts reaction.
Stereoselective one-pot, three-component synthesis of 4-aryltetrahydropyran via Prins-Friedel-Crafts reaction.
The Journal of organic chemistry (2009-02-17)
U C Reddy, S Bondalapati, Anil K Saikia
ABSTRACT
A diastereoselective one-pot, three-component Prins-Friedel-Crafts reaction was developed for the synthesis of 4-aryltetrahydropyran derivatives from the reaction of carbonyl compounds with homoallylic alcohol in the presence of arene promoted by boron trifluoride etherate.
Sigma-Aldrich
Boron trifluoride diethyl etherate, purified by redistillation, ≥46.5% BF3 basis
Sigma-Aldrich
Boron trifluoride diethyl etherate