Merck
  • In-situ fluorimetry: a powerful non-invasive diagnostic technique for natural dyes used in artefacts. Part II. Identification of orcein and indigo in Renaissance tapestries.
In-situ fluorimetry: a powerful non-invasive diagnostic technique for natural dyes used in artefacts. Part II. Identification of orcein and indigo in Renaissance tapestries.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-11-14)
C Clementi, C Miliani, A Romani, U Santamaria, F Morresi, K Mlynarska, G Favaro
ABSTRACT
In this paper, three Renaissance tapestries depicting scenes painted by Raffaello Sanzio, conserved at the Vatican Museum, were investigated using in-situ UV-Visible fluorimetric measurements. The results show that this technique is suitable for the detection of natural organic colorants used for dyeing the threads woven in these tapestries. The emission signals detected on red-purple colours were assigned to the colorant orcein and those on different nuances of blue and green colours to indigo by comparison with data from reference laboratory samples. The assignments were supported by chromatographic experiments carried out on threads taken from the back side of the tapestry in the same points analysed by spectrofluorimentry.
Sigma-Aldrich
Orcein, synthetic
Sigma-Aldrich
Orcein, synthetic, certified by the BSC