Merck
  • Lithium thiazolidine-4-carboxylate: synthesis, spectroscopic characterization and preliminary in vitro cytotoxic studies in human HeLa cells.
Lithium thiazolidine-4-carboxylate: synthesis, spectroscopic characterization and preliminary in vitro cytotoxic studies in human HeLa cells.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-03-21)
Pedro P Corbi, Fabiana C Andrade, Antonio C Massabni, Tassiele A Heinrich, Pedro P C Souza, Cláudio M Costa-Neto
ABSTRACT
A new water-soluble lithium salt of thiazolidine-4-carboxylic acid was synthesized and characterized by chemical and spectroscopic techniques. Elemental and mass spectrometric (ESI-MS) analyses of the solid compound fit to the composition LiC(4)H(6)NSO(2). (1)H, (13)C nuclear magnetic resonance (NMR), [(1)H-(15)N] NMR and infrared (IR) analyses permitted to elucidate the structure of the compound. Biological activity was evaluated by cytotoxic analysis using HeLa cells. Determination of cell death was assessed using a tetrazolium salt colorimetric assay, which reflects the cells viability.
Sigma-Aldrich
L-4-Thiazolidinecarboxylic acid, 98%