- Fragrance material review on cinnamyl isobutyrate.
Fragrance material review on cinnamyl isobutyrate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-24)
S P Bhatia, G A Wellington, J Cocchiara, J Lalko, C S Letizia, A M Api
PMID18035465
ABSTRACT
A toxicologic and dermatologic review of cinnamyl isobutyrate when used as a fragrance ingredient is presented.