Merck
  • Pentafluorophenyl copper: aggregation and complexation phenomena, photoluminescence properties, and applications as reagent in organometallic synthesis.
Pentafluorophenyl copper: aggregation and complexation phenomena, photoluminescence properties, and applications as reagent in organometallic synthesis.
Dalton transactions (Cambridge, England : 2003) (2007-07-04)
Frieder Jäkle
ABSTRACT
Recent advances in the chemistry of pentafluorophenyl copper are discussed, including the observation of strongly luminescent adducts with pyridine, the first successful structural characterization of an organocopper-arene complex, and the complexation with electron-rich transition metal complexes such as ferrocene derivatives. In addition, new applications in synthetic organometallic chemistry are discussed, which include the discovery of tin/copper exchange reactions for the preparation of organocopper compounds that are otherwise not readily accessible, and the selective transfer of the C(6)F(5) groups to boron to form catalytically active and electronically interesting organoboron polymers.
Sigma-Aldrich
Pentafluorophenol, ReagentPlus®, ≥99%