Merck
  • One-electron reduction of methanesulfonyl chloride. The fate of MeSO2Cl(.)(-) and MeSO2(.) intermediates in oxygenated solutions and their role in the cis-trans isomerization of mono-unsaturated fatty acids.
One-electron reduction of methanesulfonyl chloride. The fate of MeSO2Cl(.)(-) and MeSO2(.) intermediates in oxygenated solutions and their role in the cis-trans isomerization of mono-unsaturated fatty acids.
Journal of the American Chemical Society (2007-06-22)
Maurizio Tamba, Katalin Dajka, Carla Ferreri, Klaus-Dieter Asmus, Chryssostomos Chatgilialoglu
ABSTRACT
The one-electron reduction of methanesulfonyl chloride (MeSO2Cl) leads, in the first instance, to an electron adduct MeSO2Cl(.)(-) which lives long enough for direct detection and decays into sulfonyl radicals MeSO2(.) and Cl(-), with k = 1.5 x 10(6) s(-1). Both, MeSO2Cl(.)(-) and MeSO2(.) showed a similar absorption in the UV with lambdamax of 320 nm. In the presence of oxygen, MeSO2Cl(.)(-) transfers an electron to O(2) and establishes an equilibrium with superoxide. The rate constant for the forward reaction was measured to 4.1 x 10(9) M(-1) s(-1), while for the back reaction only an interval of 1.7 x 10(5) to 1.7 x 10(6) M(-1) s(-1) could be estimated, with a somewhat higher degree of confidence for the lower value. This corresponds to an equilibrium constant in the range of 2.4 x 10(3) to 2.4 x 10(4). With reference to E degrees (O2/O2(.)(-)) = -155 mV, the redox potential of the sulfonyl chloride couple, E degrees (MeSO2Cl/MeSO2Cl(.)(-)), thus results between being equal to -355 and -414 mV (vs NHE). MeSO2Cl(.)(-) reduces (besides O2) 4-nitroacetophenone. The underlying electron transfer took place with k = 1.5 x 10(9) M(-1) s(-1), corroborating an E degrees for the sulfonyl chloride couple significantly exceeding the above listed lower value. The MeSO2(.) radical added to oxygen with a rate constant of 1.1 x 10(9) M(-1) s(-1). Re-dissociation of O2 from MeSO2OO(.) occurred only very slowly, if at all, that is, with k < 10(5) s(-1). MeSO2(.) radicals can act as the catalyst for the cis-trans isomerization of several Z- and E-mono-unsaturated fatty acid methyl esters in homogeneous solution. The effectiveness of the isomerization processes has been addressed, and in the presence of oxygen the isomerization is completely suppressed.
Sigma-Aldrich
Methanesulfonyl chloride, ≥99.7%
Sigma-Aldrich
Methanesulfonyl chloride, purum, ≥98.0% (AT)