Merck
  • Diethylhexylphthalate (DEHP) and triethylhexyltrimellitate (TEHT) PVC plasticizers assay in blood.
Diethylhexylphthalate (DEHP) and triethylhexyltrimellitate (TEHT) PVC plasticizers assay in blood.
Bollettino chimico farmaceutico (1991-06-01)
P M Boselli, E Cislaghi
ABSTRACT
The interest in the PVC biocompatibility of the apparatus used both for dialysis and any other extracorporeal blood circulation is so real that a method to determine PVC plasticizer concentration in blood is properly needed. The described method might be easier than other ones we are allowed to use at the moment.
Sigma-Aldrich
Trioctyl trimellitate, 99%