Merck
  • Ammonium hexafluorosilicate elicits calcium phosphate precipitation and shows continuous dentin tubule occlusion.
Ammonium hexafluorosilicate elicits calcium phosphate precipitation and shows continuous dentin tubule occlusion.
Dental materials : official publication of the Academy of Dental Materials (2007-06-05)
Toshiyuki Suge, Akiko Kawasaki, Kunio Ishikawa, Takashi Matsuo, Shigeyuki Ebisu
ABSTRACT
Diamine silver fluoride [AgF: (NH(3))(2)AgF] has been used clinically in Japan, as it reduces dental caries and dentin hypersensitivity. However, AgF stains the teeth black due to silver precipitation. To overcome this drawback, the authors prepared ammonium hexafluorosilicate [SiF: (NH(4))(2)SiF(6)], which does not stain the teeth, and SiF occluded open dentin tubules completely with silica-calcium phosphate precipitate. The aim of this study was to evaluate the duration of dentin tubule occlusion after SiF treatment in a simulated oral environment. To simulate dentin tubules subject to dentin hypersensitivity, dentin disks were treated with EDTA for 2 min. The disks were treated with 0.476 mol/L SiF for 3 min, and then the disks were immersed in synthetic saliva, which was regularly replenished to maintain its ionic concentration, for up to 7 days. The occluding ability of the dentin tubules was evaluated using scanning electron microscopy (SEM), and the hydraulic conductance was measured following Pashley's method at regular intervals. SEM photographs demonstrated that dentin tubules were occluded homogeneously and completely with the precipitate at 7 days after treatment with SiF. In addition, newly formed calcium phosphate precipitate was present at the dentin surface. The dentin permeability showed a consistently low value throughout the experimental period. The values immediately after SiF treatment and 7 days after immersion were 11.9+/-3.7% and 7.9+/-2.9%, respectively. Ammonium hexafluorosilicate is useful for the treatment of dentin hypersensitivity, since ammonium hexafluorosilicate induced calcium phosphate precipitation from the saliva; therefore, it has a continuous effect on dentin tubules occlusion under a simulated oral environment.
Product Description
Supelco
Potassium hexafluorosilicate, ≥99.0% (T)