Merck
[Constituents of Liguliria vellerea (Franch.) Hand-Mazz].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2003-06-05)
Y S Li, S D Luo, M Zhang, J J Chen, Z T Wang
ABSTRACT
To study the chemical constituents of Ligularia vellerea. The compounds were isolated by column chromatography, and the structures were identified by NMR spectral data and other methods. Seven compounds were isolated and identified as 4-hydroxyacetophenone, 8 alpha-hydroxy-7(11)-eremophilen-12, 8 beta-olide, umbelliferone, tiglic acid, 6 beta-hydroxy-7(11)-eremophilen-12, 8 alpha-olide, daucosterin, beta-sitosterol and stigmasterol. All the compounds were isolated for the first time from this plant.
Sigma-Aldrich
trans-2-Methyl-2-butenoic acid, ≥99%, FG
Sigma-Aldrich
trans-2,3-Dimethylacrylic acid, 98%