Merck
  • Respiratory tract responses to repeated inhalation of an oxidant and acid gas-particle air pollutant mixture.
Respiratory tract responses to repeated inhalation of an oxidant and acid gas-particle air pollutant mixture.
Toxicological sciences : an official journal of the Society of Toxicology (2001-05-16)
W J Mautz, M T Kleinman, D K Bhalla, R F Phalen
ABSTRACT
The purpose of this study was to examine a broad range of toxicologic responses in rats exposed to a multi-component pollutant atmosphere. Cumulative and adaptive respiratory tract responses to 3 concentrations of an inhaled particle-oxidant mixture were examined in Fisher 344 N rats exposed 4 h/day, 3 days/week for 4 weeks. The mixtures contained O3, NO2, NH4HSO4, carbon particles, and HNO3 vapor. Irritant-induced, rapid-shallow breathing responses were present during the first 4-h exposure to medium and high concentrations. Successive exposures showed diminished responses in medium concentrations and exacerbated responses in high concentrations. At the end of 4 weeks, rats exposed to high concentrations exhibited lung lesions. Lavaged pulmonary macrophages showed dose-dependent depressions of Fc-receptor binding and phagocytosis. Lung tissue macrophages showed dose-dependent increases in acid phosphatase staining density and carbon particles. Respiratory tract clearance of tracer particles was not significantly affected by the exposures. Broncho-alveolar epithelial permeability was increased by the high concentration. Epithelial cell-proliferation labeling showed a dose-dependent increase at all levels of the respiratory tract. Progressively exacerbated breathing-pattern responses at high concentrations were associated with lung lesions and high cell-proliferation labeling in the nose transitional epithelium and terminal bronchioles. Attenuating or adaptive breathing-pattern responses occurred in the presence of smaller, but in many cases still significant, compromise of respiratory functions. Either attenuating or exacerbated breathing-pattern responses can occur in the presence of a significant dose-dependent compromise of other respiratory functions and lung tissue injury.
Sigma-Aldrich
Ammonium hydrogensulfate, 99.99% trace metals basis