Merck
  • Carbon-based quantitation of pyrethrins by supercritical-fluid chromatography.
Carbon-based quantitation of pyrethrins by supercritical-fluid chromatography.
Journal of biochemical and biophysical methods (2000-06-28)
B Wenclawiak, A Otterbach
ABSTRACT
All six insecticide active ingredients in pyrethrum extract were quantified by supercritical fluid chromatography and carbon calibration. Allethrin is a suitable reference compound for carbon calibration and pyrethrins calibrations. Carbon quantification in SFC is also applied to pyrethroids (phenothrin, permethrin, cypermethrin, fenvalerate and deltamethrin) and alkanes. Halogen substitution on pyrethroids requires halogens on the reference calibration compound. The method was applied to commercial extracts.
Product Description
Supelco
Eicosane, analytical standard
Sigma-Aldrich
Eicosane, 99%