- PD 142893, SB 209670, and BQ 788 selectively antagonize vascular endothelial versus vascular smooth muscle ET(B)-receptor activity in the rat.
PD 142893, SB 209670, and BQ 788 selectively antagonize vascular endothelial versus vascular smooth muscle ET(B)-receptor activity in the rat.
The purpose of this study was to determine whether vascular endothelial and vascular smooth-muscle endothelin ET(B) receptors could be quantitatively differentiated by PD 142893 (PD), SB 209670 (SB), and BQ 788 (BQ) in the same species by using closely matched experimental conditions. The isolated perfused rat kidney (vascular smooth muscle) and isolated perfused rat mesentery (vascular endothelium) were challenged with increasing bolus doses of sarafotoxin S6c in the absence and presence of antagonist. PD, SB, and BQ produced parallel concentration-dependent rightward shifts in the S6c dose-response curve in the kidney. PD and SB also produced parallel concentration-dependent rightward shifts in the S6c dose-response curve in the mesentery. In contrast, BQ produced an insurmountable antagonism. Schild-derived pA2 values for PD and SB were significantly greater for inhibiting endothelial versus smooth-muscle ET(B) receptors. Furthermore, PD and SB differed in their relative potency between the two assays. Because BQ produced an insurmountable antagonism in the mesentery, it was not possible quantitatively to compare the antagonist activity in the two assays. These results indicate that PD, SB, and BQ selectively antagonize endothelial ET(B)-receptor activity over smooth-muscle ET(B)-receptor activity.