Merck
  • Rehmannia glutinosa polysaccharide induces maturation of murine bone marrow derived Dendritic cells (BMDCs).
Rehmannia glutinosa polysaccharide induces maturation of murine bone marrow derived Dendritic cells (BMDCs).
International journal of biological macromolecules (2012-12-19)
Zhenjie Zhang, Yiming Meng, Yanxia Guo, Xin He, Qingrui Liu, Xiufeng Wang, Fengping Shan
ABSTRACT
Purified Rehmannia glutinosa polysaccharide (RGP) is used as functional foods for the prevention and treatment of various diseases. In this study, we examined the effects of RGP on phenotypic and functional maturation of murine bone marrow derived Dendritic cells (BMDCs). Phenotypic maturation of BMDCs was confirmed by conventional scanning electron microscopy (SEM), flow cytometry (FCM) and functional maturation by transmission electron microscopy (TEM), cytochemistry assay, Acid phosphatase (ACP) activity, FITC-dextran, bio-assay and enzyme linked immunosorbent assay (ELISA).We found that RGP up-regulated the expression of CD40, CD80, CD83, CD86 and MHC II molecules of BMDCs, down-regulated pinocytosis and phagocytosis activity, induced IL-12 and TNF-α production of BMDCs. It is therefore concluded that RGP can effectively promote the maturation of DCs. Our study provides evidence and rationale on using RGP in various clinical conditions to enhance host immunity and suggests RGP as a potent adjuvant for the design of DC-based vaccines.
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid