- Diagnosis and classification of hemolytic uremic syndrome: the Hungarian experience.
Diagnosis and classification of hemolytic uremic syndrome: the Hungarian experience.
Hemolytic uremic syndrome (HUS) is a rare disease with various etiologies, making the identification of the specific forms and appropriate treatment difficult. Therefore, clinical and laboratory data from these patients need to be analyzed in national and international registries. Herein we have described 47 Hungarian HUS patients with detailed laboratory and clinical data obtained between 2008 and 2010. Blood samples and clinical data of 47 patients with HUS diagnosed according to characteristic clinical signs were submitted for diagnostic evaluation, including complement protein and genetic analysis, measurement of ADAMTS13 activity and antibody analysis against O157LPS and factor H. There were 8 patients with typical diarrhea-positive HUS; 13 with atypical HUS (aHUS) and 26 with secondary HUS/thrombotic thrombocytopenic purpura group characterized by signs of complement consumption and decreased ADAMTS13 activity. Thus, decreased total alternative pathway activity is a promising diagnostic parameter with good sensitivity for aHUS. These observations highlight the requirement for multiple diagnostic tests together with clinical data to identify the specific cause of HUS. Because the long-term prognosis of aHUS, eg, graft survival after renal transplantation, may vary according to the molecular etiology, it is important for all affected patients to undergo a detailed molecular diagnosis of the disease. There is a clear clinical need for the development and application of novel assay in this field to allow more rapid efficient diagnosis of patients who undergo a first episode of HUS.