Merck
  • Monitoring the hydrolysis of toxic organophosphonate nerve agents in aqueous buffer and in bicontinuous microemulsions by use of diisopropyl fluorophosphatase (DFPase) with (1)H- (31)P HSQC NMR spectroscopy.
Monitoring the hydrolysis of toxic organophosphonate nerve agents in aqueous buffer and in bicontinuous microemulsions by use of diisopropyl fluorophosphatase (DFPase) with (1)H- (31)P HSQC NMR spectroscopy.
Analytical and bioanalytical chemistry (2009-11-28)
Jürgen Gäb, Marco Melzer, Kai Kehe, Stefan Wellert, Thomas Hellweg, Marc-Michael Blum
ABSTRACT
The enzyme diisopropyl fluorophosphatase (DFPase, EC 3.1.8.2) from the squid Loligo vulgaris effectively catalyzes the hydrolysis of diisopropyl fluorophosphate (DFP) and a number of organophosphorus nerve agents, including sarin, soman, cyclosarin, and tabun. Until now, determination of kinetic data has been achieved by use of techniques such as pH-stat titration, ion-selective electrodes, and a recently introduced method based on in situ Fourier-transform infrared (FTIR) spectroscopy. We report the use of 1D (1)H-(31)P HSQC NMR spectroscopy as a new method for real-time quantification of the hydrolysis of toxic organophosphonates by DFPase. The method is demonstrated for the agents sarin (GB), soman (GD), and cyclosarin (GD) but can also be used for V-type nerve agents, for example VX. Besides buffered aqueous solutions the method was used to determine enzymatic activities in a biodiesel-based bicontinuous microemulsion that serves as an example of complex decontamination media, for which other established techniques often fail. The method is non-invasive and requires only limited manual handling of small volumes of liquid (700 microL), which adds to work safety when handling highly toxic organophosphorus compounds. Limits of detection are slightly below 100 micromol L(-1) on a 400 MHz spectrometer with 16 FIDs added for a single time frame. The method is not restricted to DFPase but can be used with other phosphotriesterases, for example paraxonase (PON), and even reactive chemicals, for example oximes and other nucleophiles, as long as the reaction components are compatible with the NMR experiment.
Sigma-Aldrich
Diisopropyl-fluorophosphatase, ≥30 U/mg