- In vivo processing of nonanchored Yapsin 1 (Yap3p).
In vivo processing of nonanchored Yapsin 1 (Yap3p).
A C-terminally truncated form of yapsin 1 (yeast aspartic protease 3) was overexpressed in yeast and its processing through the secretory pathway was followed by pulse-labeling and immunoprecipitation studies. In the soluble cell extract, three forms of yapsin 1-87, 74, and 18 kDa-were found. Identification of these forms of yapsin 1 using different antisera suggests that the 87-kDa form is pro-yapsin 1, which is processed into two subunits, alpha (18 kDa) and beta (74 kDa), by cleavage at a loop region not found in traditional aspartic proteases. By use of a temperature-sensitive mutant strain, sec18, the generation of the two subunits was found to occur in the endoplasmic reticulum. An active site-mutated yapsin 1 was not processed into the two subunits, suggesting that this process occurs in an autocatalytic manner.