- Effect of oxidative stress on plasma membrane fluidity of THP-1 induced macrophages.
Effect of oxidative stress on plasma membrane fluidity of THP-1 induced macrophages.
Plasma membrane is one of the preferential targets of reactive oxygen species which cause lipid peroxidation. This process modifies membrane properties such as membrane fluidity, a very important physical feature known to modulate membrane protein localization and function. The aim of this study is to evaluate the effect of oxidative stress on plasma membrane fluidity regionalization of single living THP-1 macrophages. These cells were oxidized with H(2)O(2) at different concentrations, and plasma membrane fluidity was analyzed by two-photon microscopy in combination with the environment-sensitive probe Laurdan. Results show a significant H(2)O(2) concentration dependent increase in the frequency of rigid lipid regions, mainly attributable to lipid rafts, at the expense of the intermediate fluidity regions. A novel statistical analysis evaluated changes in size and number of lipid raft domains under oxidative stress conditions, as lipid rafts are platforms aiding cell signaling and are thought to have relevant roles in macrophage functions. It is shown that H(2)O(2) causes an increase in the number, but not the size, of raft domains. As macrophages are highly resistant to H(2)O(2), these new raft domains might be involved in cell survival pathways.