Merck
  • Prolonged hypernutrition impairs TREM2-dependent efferocytosis to license chronic liver inflammation and NASH development.
Prolonged hypernutrition impairs TREM2-dependent efferocytosis to license chronic liver inflammation and NASH development.
Immunity (2022-12-16)
Xiaochen Wang, Qifeng He, Chuanli Zhou, Yueyuan Xu, Danhui Liu, Naoto Fujiwara, Naoto Kubota, Arielle Click, Polly Henderson, Janiece Vancil, Cesia Ammi Marquez, Ganesh Gunasekaran, Myron E Schwartz, Parissa Tabrizian, Umut Sarpel, Maria Isabel Fiel, Yarui Diao, Beicheng Sun, Yujin Hoshida, Shuang Liang, Zhenyu Zhong
ABSTRACT
Obesity-induced chronic liver inflammation is a hallmark of nonalcoholic steatohepatitis (NASH)-an aggressive form of nonalcoholic fatty liver disease. However, it remains unclear how such a low-grade, yet persistent, inflammation is sustained in the liver. Here, we show that the macrophage phagocytic receptor TREM2, induced by hepatocyte-derived sphingosine-1-phosphate, was required for efferocytosis of lipid-laden apoptotic hepatocytes and thereby maintained liver immune homeostasis. However, prolonged hypernutrition led to the production of proinflammatory cytokines TNF and IL-1β in the liver to induce TREM2 shedding through ADAM17-dependent proteolytic cleavage. Loss of TREM2 resulted in aberrant accumulation of dying hepatocytes, thereby further augmenting proinflammatory cytokine production. This ultimately precipitated a vicious cycle that licensed chronic inflammation to drive simple steatosis transition to NASH. Therefore, impaired macrophage efferocytosis is a previously unrecognized key pathogenic event that enables chronic liver inflammation in obesity. Blocking TREM2 cleavage to restore efferocytosis may represent an effective strategy to treat NASH.
Sigma-Aldrich
CYM-5442, ≥98% (HPLC)