- FSTL3 is associated with prognosis and immune cell infiltration in lung adenocarcinoma.
FSTL3 is associated with prognosis and immune cell infiltration in lung adenocarcinoma.
FSTL3 expression is altered in various types of cancer. However, the role and mechanism of action of FSTL3 in lung adenocarcinoma development and tumor immunity are unknown. We investigated the association between FSTL3 expression and clinical characteristics and immune cell infiltration in lung adenocarcinoma samples from The Cancer Genome Atlas (TCGA) and a separate validation set from our hospital. Data on immune system infiltration, gene expression, and relevant clinical information were obtained by analyzing lung adenocarcinoma sample data from TCGA database. Using online tools like GEPIA, the correlations between FSTL3 expression and prognosis, clinical stage, survival status, and tumor-infiltrating immune cells were examined. In a validation dataset, immunohistochemistry was performed to analyze FSTL3 expression and its related clinical characteristics. FSTL3 expression was markedly reduced in patients with lung adenocarcinoma. N stage, pathological stage, and overall survival were significantly correlated with FSTL3 expression. According to GSEA, FSTL3 is strongly linked to signaling pathways such as DNA replication and those involved in cell cycle regulation. Examination of TCGA database and TIMER online revealed a correlation between FSTL3 and B cell, T cell, NK cell, and neutrophil levels. The prognosis of patients with lung adenocarcinoma was significantly affected by six genes (KRT6A, VEGFC, KRT14, KRT17, SNORA12, and KRT81) related to FSTL3. FSTL3 is significantly associated with the prognosis and progression of lung adenocarcinoma and the infiltration of immune cells. Thus, targeting FSTL3 and its associated genes in immunotherapy could be potentially beneficial for the treatment of lung adenocarcinoma.