- Investigating the Electric Field Lysis of Exosomes Immobilized on the Screen-Printed Electrode and Electrochemical Sensing of the Lysed-Exosome-Derived Protein.
Investigating the Electric Field Lysis of Exosomes Immobilized on the Screen-Printed Electrode and Electrochemical Sensing of the Lysed-Exosome-Derived Protein.
It is important to isolate exosomes (<150 nm) from biofluid for diagnosis or prognosis purposes, followed by sensing of exosomal proteins. In the present work, exosomes are isolated from human serum by immobilizing on a Screen-Printed Electrode (SPE) followed by electric field lysis and electrochemical impedance spectroscopy (EIS)-based sensing of relevant exosomal proteins (HSP70 and HER2). Upon immobilization of exosomes on the surface, the role of different electrical signals (sinusoidal and square wave) in the lysis of exosomes was studied by varying the frequency and voltage. HSP70 was used for EIS to determine the optimal voltage and frequency for lysing the exosomes. It was observed that the low frequencies and, specifically, sinusoidal signals are ideal for lysing exosomes as compared to square signals. The relative quantity of HSP70 obtained by lysing with different voltages (sinusoidal waveform) was compared using Western blotting. After electric field lysis of the exosome with an optimized signal, HER2, a breast cancer biomarker, was detected successfully from serum by EIS. In the proposed technique, 3.5 × 108 exosomes/mL were isolated from serum. With the limit of detection of 10 pg, the designed cell showed a linear detection of HER2 from 0.1 ng to 1 µg. It was observed from the results that the electric field lysis of exosomes not only plays a significant role in releasing the cargo protein but also improves the sensing of surface proteins associated with exosomes.