Merck
  • Experimental Support for Human Papillomavirus Genome Amplification Early after Infectious Delivery.
Experimental Support for Human Papillomavirus Genome Amplification Early after Infectious Delivery.
Journal of virology (2023-05-24)
Katarzyna Zwolinska, Malgorzata Bienkowska-Haba, Rona S Scott, Timothy Keiffer, Martin Sapp
ABSTRACT
Even though replication and transcription of human papillomavirus type 16 (HPV16) has been intensively studied, little is known about immediate-early events of the viral life cycle due to the lack of an efficient infection model allowing genetic dissection of viral factors. We employed the recently developed infection model (Bienkowska-Haba M, Luszczek W, Myers JE, Keiffer TR, et al. 2018. PLoS Pathog 14:e1006846) to investigate genome amplification and transcription immediately after infectious delivery of viral genome to nuclei of primary keratinocytes. Using 5-ethynyl-2'-deoxyuridine (EdU) pulse-labeling and highly sensitive fluorescence in situ hybridization, we observed that the HPV16 genome is replicated and amplified in an E1- and E2-dependent manner. Knockout of E1 resulted in failure of the viral genome to replicate and amplify. In contrast, knockout of the E8^E2 repressor led to increased viral genome copy number, confirming previous reports. Genome copy control by E8^E2 was confirmed for differentiation-induced genome amplification. Lack of functional E1 had no effect on transcription from the early promoter, suggesting that viral genome replication is not required for p97 promoter activity. However, infection with an HPV16 mutant virus defective for E2 transcriptional function revealed a requirement of E2 for efficient transcription from the early promoter. In the absence of the E8^E2 protein, early transcript levels are unaltered and even decreased when normalized to genome copy number. Surprisingly, a lack of functional E8^E2 repressor did not affect E8^E2 transcript levels when normalized to genome copy number. These data suggest that the main function of E8^E2 in the viral life cycle is to control genome copy number. IMPORTANCE It is being assumed that human papillomavirus (HPV) utilizes three different modes of replication during its life cycle: initial amplification during the establishment phase, genome maintenance, and differentiation-induced amplification. However, HPV16 initial amplification was never formally proven due to a lack of an infection model. Using our recently established infection model (Bienkowska-Haba M, Luszczek W, Myers JE, Keiffer TR, et al. 2018. PLoS Pathog 14:e1006846), we demonstrate herein that viral genome is indeed amplified in an E1- and E2-dependent manner. Furthermore, we find that the main function of the viral repressor E8^E2 is to control viral genome copy number. We did not find evidence that it regulates its own promoter in a negative feedback loop. Our data also suggest that the E2 transactivator function is required for stimulation of early promoter activity, which has been debated in the literature. Overall, this report confirms the usefulness of the infection model for studying early events of the HPV life cycle using mutational approaches.
Sigma-Aldrich
UM-SCC-47 HPV-16 Positive Squamous Carcinoma Cell Line, suitable in vitro model of H&N carcinoma studies