Merck
  • Potential application of let-7a antagomir in injured peripheral nerve regeneration.
Potential application of let-7a antagomir in injured peripheral nerve regeneration.
Neural regeneration research (2022-12-27)
Qian-Qian Chen, Qian-Yan Liu, Pan Wang, Tian-Mei Qian, Xing-Hui Wang, Sheng Yi, Shi-Ying Li
ABSTRACT
Neurotrophic factors, particularly nerve growth factor, enhance neuronal regeneration. However, the in vivo applications of nerve growth factor are largely limited by its intrinsic disadvantages, such as its short biological half-life, its contribution to pain response, and its inability to cross the blood-brain barrier. Considering that let-7 (human miRNA) targets and regulates nerve growth factor, and that let-7 is a core regulator in peripheral nerve regeneration, we evaluated the possibilities of let-7 application in nerve repair. In this study, anti-let-7a was identified as the most suitable let-7 family molecule by analyses of endogenous expression and regulatory relationship, and functional screening. Let-7a antagomir demonstrated biosafety based on the results of in vivo safety assessments and it entered into the main cell types of the sciatic nerve, including Schwann cells, fibroblasts and macrophages. Use of hydrogel effectively achieved controlled, localized, and sustained delivery of let-7a antagomir. Finally, let-7a antagomir was integrated into chitosan conduit to construct a chitosan-hydrogel scaffold tissue-engineered nerve graft, which promoted nerve regeneration and functional recovery in a rat model of sciatic nerve transection. Our study provides an experimental basis for potential in vivo application of let-7a.
Sigma-Aldrich
Monoclonal Anti-Thy 1.1 antibody produced in mouse, clone TN-26, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Anti-Neurofilament 160/200 antibody, Mouse monoclonal, ~2 mg/mL, clone RMdO20, purified from hybridoma cell culture