Merck
  • Omicron's binding to sotrovimab, casirivimab, imdevimab, CR3022, and sera from previously infected or vaccinated individuals.
Omicron's binding to sotrovimab, casirivimab, imdevimab, CR3022, and sera from previously infected or vaccinated individuals.
iScience (2022-03-22)
Anna-Lena Mader, Leonid Tydykov, Vivian Glück, Manuela Bertok, Tanja Weidlich, Christine Gottwald, Alexa Stefl, Matthias Vogel, Annelie Plentz, Josef Köstler, Bernd Salzberger, Jürgen J Wenzel, Hans Helmut Niller, Jonathan Jantsch, Ralf Wagner, Barbara Schmidt, Thomas Glück, André Gessner, David Peterhoff
ABSTRACT
SARS-CoV-2 Omicron is the first pandemic variant of concern exhibiting an abrupt accumulation of mutations particularly in the receptor-binding domain that is a critical target of vaccination induced and therapeutic antibodies. Omicron's mutations did only marginally affect the binding of ACE2, and the two antibodies Sotrovimab and CR3022 but strongly impaired the binding of Casirivimab and Imdevimab. Moreover, as compared with Wuhan, there is reduced serum reactivity and a pronounced loss of competitive surrogate virus neutralization (sVN) against Omicron in naïve vaccinees and in COVID-19 convalescents after infection and subsequent vaccination. Finally, although the booster vaccination response conferred higher titers and better sVN, the effect was nonetheless significantly lower compared with responses against Wuhan. Overall, our data suggest that the antigenicity of Omicrons receptor binding motive has largely changed but antibodies such as Sotrovimab targeting other conserved sites maintain binding and therefore hold potential in prophylaxis and treatment of Omicron-induced COVID-19.
Product Description
Roche
Streptavidin-POD conjugate, liquid, pkg of 1 mL
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)