Merck
  • miR-144-3p ameliorates the progression of osteoarthritis by targeting IL-1β: Potential therapeutic implications.
miR-144-3p ameliorates the progression of osteoarthritis by targeting IL-1β: Potential therapeutic implications.
Journal of cellular physiology (2021-03-28)
Yen-You Lin, Chih-Yuan Ko, Shan-Chi Liu, Yu-Han Wang, Chin-Jung Hsu, Chun-Hao Tsai, Tsung-Ju Wu, Chih-Hsin Tang
ABSTRACT
The pro-inflammatory cytokine interleukin 1 beta (IL-1β) plays a critical role in osteoarthritis (OA) disease pathogenesis. MicroRNA (miRNA) activity is related to inflammation in OA and some miRNAs specifically regulate IL-mediated degradation of cartilage type II collagen. Previous studies have indicated that miR-144-3p is a useful target in the regulation of pro-inflammatory cytokines in different diseases. However, the role of miR-144-3p in OA is unclear. In this study, we observed a negative correlation between miR-144-3p and IL-1β expression in OA. miR-144-3p mimic transfection of OA synovial fibroblasts downregulated levels of IL-1β expression, while blocking the MAPK, PI3K/Akt, and NF-κB signaling pathways relating to IL-1β production, and effectively increased miR-144-3p expression in OASFs. Findings from an anterior cruciate ligament transection rat model revealed that administration of miR-144-3p mimic effectively ameliorated OA progression and reduced the numbers of IL-1β-positive cells in synovial tissue. This study suggests that miR-144-3p is a useful therapeutic target in OA disease.
Sigma-Aldrich
Akt1/2 kinase inhibitor, ≥98% (HPLC)
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%