Merck
  • The effect of statins and the synthetic LXR agonist T0901317 on expression of ABCA1 transporter protein in human lung epithelial cell lines in vitro.
The effect of statins and the synthetic LXR agonist T0901317 on expression of ABCA1 transporter protein in human lung epithelial cell lines in vitro.
Pharmacological reports : PR (2019-11-02)
Patrick He, Aaron Smith, Ingrid C Gelissen, Alaina J Ammit
ABSTRACT
The pathogenesis of chronic obstructive pulmonary disease (COPD) is associated with dyslipidemia, an established co-morbidity. Statins treat hypercholesterolemia, but more recently have been trailed in the setting of COPD for their potential anti-inflammatory benefits. The outcomes of prospective trials however have been inconsistent. Thus, we hypothesize that the variation in results may have been due to statin-induced downregulation of ATP-binding cassette transporter A1 (ABCA1), thereby reducing cholesterol export. This study aims to elucidate whether statin treatment in a cellular model of COPD leads to a decrease in ABCA1 protein expression. To mimic the inflammatory environment of COPD, two commonly used lung epithelial cell lines (BEAS-2B and A549) were treated with tumor necrosis factor (TNF), and co-treated with cholesterol/25-hydroxycholesterol (25-OH) to mimic dyslipidemia. ABCA1 protein was detected by Western Blotting. We unexpectedly showed that statins did not affect ABCA1 expression. However, the LXR agonist T0901317 significantly increased ABCA1 expression in both cell lines, while TNF, cholesterol or 25-OH induced ABCA1 protein upregulation in BEAS-2B cells, indicating cell line differences in response. There was also evidence of synergistic impacts of combined treatments on ABCA1 upregulation in BEAS-2B cells. Statins did not have an impact on ABCA1 expression in lung epithelial cell lines, disproving our original hypothesis. However, we showed for the first time, the effect of the inflammatory cytokine TNF, cholesterol/25-OH, statins and the LXR agonist T0901317 on expression of ABCA1 transporter protein in human lung epithelial cell lines in vitro. We hope that these in vitro studies may prove beneficial for addressing dyslipidemia in COPD in the future.
Sigma-Aldrich
Monoclonal Anti-Heat Shock Protein 70 antibody produced in mouse, clone BRM-22, ascites fluid
Sigma-Aldrich
Anti-ABCA1 Antibody, clone AB.H10, clone AB.H10, Chemicon®, from mouse