Merck
  • Slow development of bladder malfunction parallels spinal cord fiber sprouting and interneurons' loss after spinal cord transection.
Slow development of bladder malfunction parallels spinal cord fiber sprouting and interneurons' loss after spinal cord transection.
Experimental neurology (2021-11-27)
Andrea M Sartori, Anna-Sophie Hofer, Myriam I Scheuber, Ruslan Rust, Thomas M Kessler, Martin E Schwab
ABSTRACT
Neurogenic lower urinary tract dysfunction typically develops after spinal cord injury. We investigated the time course and the anatomical changes in the spinal cord that may be causing lower urinary tract symptoms following injury. Rats were implanted with a bladder catheter and external urethral sphincter electromyography electrodes. Animals underwent a large, incomplete spinal transection at the T8/9 spinal level. At 1, 2-3, and 4 weeks after injury, the animals underwent urodynamic investigations. Urodynamic investigations showed detrusor overactivity and detrusor-sphincter-dyssynergia appearing over time at 3-4 weeks after injury. Lower urinary tract dysfunction was accompanied by an increase in density of C-fiber afferents in the lumbosacral dorsal horn. CRF-positive Barrington's and 5-HT-positive bulbospinal projections drastically decreased after injury, with partial compensation for the CRF fibers at 3-4 weeks. Interestingly, a decrease over time was observed in the number of GABAergic neurons in the lumbosacral dorsal horn and lamina X, and a decrease of glutamatergic cells in the dorsal horn. Detrusor overactivity and detrusor-sphincter-dyssynergia might therefore arise from a discrepancy in inhibitory/excitatory interneuron activity in the lumbosacral cord as well as input changes which develop over time after injury. The processes point to spinal plastic changes leading to malfunction of the important physiological pathway of lower urinary tract control.
Sigma-Aldrich
Anti-Calcitonin Gene Related Peptide Antibody, serum, Chemicon®