Merck
  • Homozygous SPAG6 variants can induce nonsyndromic asthenoteratozoospermia with severe MMAF.
Homozygous SPAG6 variants can induce nonsyndromic asthenoteratozoospermia with severe MMAF.
Reproductive biology and endocrinology : RB&E (2022-03-03)
Chuan Xu, Dongdong Tang, Zhongmei Shao, Hao Geng, Yang Gao, Kuokuo Li, Qing Tan, Guanxiong Wang, Chao Wang, Huan Wu, Guanjian Li, Mingrong Lv, Xiaojin He, Yunxia Cao
ABSTRACT
Multiple morphological abnormalities of the sperm flagella (MMAF) is a subtype of severe asthenoteratozoospermia with poorly understood genetic etiology. SPAG6 is a core axonemal component that plays a critical role in the formation of cilia and sperm flagella. Previous studies have reported that mutations in SPAG6 cause primary ciliary dyskinesia (PCD), but the association between SPAG6 gene variants and the MMAF phenotype has not yet been described. We performed whole-exome sequencing (WES) in two unrelated Han Chinese men with MMAF. Sanger sequencing was used to validate the candidate variants. Routine semen analysis was carried out according to the WHO guidelines (5th Edition). Sperm morphology was assessed using modified Papanicolaou staining. Scanning and transmission electron microscopy (S/TEM) was performed to observe the ultrastructural defects of the sperm flagella. Western blot analysis and immunofluorescence (IF) of spermatozoa were performed to examine the expression of SPAG6 protein. Assisted fertilization with intracytoplasmic sperm injection (ICSI) was applied. Two homozygous SPAG6 variants were identified by WES and Sanger validation in two patients with MMAF phenotype (F1 II-1: c.308C > A, p. A103D; F2 II-1: c. 585delA, p. K196Sfs*6). Semen analysis showed progressive rates of less than 1%, and most of the spermatozoa presented MMAF by Papanicolaou staining. TEM revealed that the overall axonemal ultrastructure was disrupted and primarily presented an abnormal "9 + 0" configuration. No other PCD-related symptoms were found on physical examination and medical consultations, as well as lung CT screening. The level of SPAG6 protein was significantly decreased in the spermatozoa, and IF analysis revealed that SPAG6 staining was extremely weak and discontinuous in the sperm flagella of the two patients. Notably, F1 II-1 and his wife conceived successfully after undergoing ICSI. Our research provides new evidence for a potential correlation between SPAG6 variants and the MMAF phenotype.
Sigma-Aldrich
Anti-SPAG6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution