Merck
  • IL-9 governs allergen-induced mast cell numbers in the lung and chronic remodeling of the airways.
IL-9 governs allergen-induced mast cell numbers in the lung and chronic remodeling of the airways.
American journal of respiratory and critical care medicine (2010-10-26)
Jennifer Kearley, Jonas S Erjefalt, Cecilia Andersson, Ebony Benjamin, Carla P Jones, Annette Robichaud, Sophie Pegorier, Yambasu Brewah, Timothy J Burwell, Leif Bjermer, Peter A Kiener, Roland Kolbeck, Clare M Lloyd, Anthony J Coyle, Alison A Humbles
ABSTRACT
IL-9 is a pleiotropic cytokine that has multiple effects on structural as well as numerous hematopoietic cells, which are central to the pathogenesis of asthma. The contribution of IL-9 to asthma pathogenesis has thus far been unclear, due to conflicting reports in the literature. These earlier studies focused on the role of IL-9 in acute inflammatory models; here we have investigated the effects of IL-9 blockade during chronic allergic inflammation. Mice were exposed to either prolonged ovalbumin or house dust mite allergen challenge to induce chronic inflammation and airway remodeling. We found that IL-9 governs allergen-induced mast cell (MC) numbers in the lung and has pronounced effects on chronic allergic inflammation. Anti-IL-9 antibody-treated mice were protected from airway remodeling with a concomitant reduction in mature MC numbers and activation, in addition to decreased expression of the profibrotic mediators transforming growth factor-β1, vascular endothelial growth factor, and fibroblast growth factor-2 in the lung. Airway remodeling was associated with impaired lung function in the peripheral airways and this was reversed by IL-9 neutralization. In human asthmatic lung tissue, we identified MCs as the main IL-9 receptor expressing population and found them to be sources of vascular endothelial growth factor and fibroblast growth factor-2. Our data suggest an important role for an IL-9-MC axis in the pathology associated with chronic asthma and demonstrate that an impact on this axis could lead to a reduction in chronic inflammation and improved lung function in patients with asthma.