Merck
  • PHD finger protein 20-like protein 1 (PHF20L1) in ovarian cancer: from its overexpression in tissue to its upregulation by the ascites microenvironment.
PHD finger protein 20-like protein 1 (PHF20L1) in ovarian cancer: from its overexpression in tissue to its upregulation by the ascites microenvironment.
Cancer cell international (2022-01-08)
Dulce Rosario Alberto-Aguilar, Verónica Ivonne Hernández-Ramírez, Juan Carlos Osorio-Trujillo, Dolores Gallardo-Rincón, Alfredo Toledo-Leyva, Patricia Talamás-Rohana
ABSTRACT
Ovarian cancer is the most aggressive gynecological malignancy. Transcriptional regulators impact the tumor phenotype and, consequently, clinical progression and response to therapy. PHD finger protein 20-like protein 1 (PHF20L1) is a transcriptional regulator with several isoforms, and studies on its role in ovarian cancer are limited. We previously reported that PHF20L1 is expressed as a fucosylated protein in SKOV-3 cells stimulated with ascites from patients with ovarian cancer. We decided to analyze the expression of PHF20L1 in ovarian cancer tissues, determine whether a correlation exists between PHF20L1 expression and patient clinical data, and analyze whether ascites can modulate the different isoforms of this protein. Ovarian cancer biopsies from 29 different patients were analyzed by immunohistochemistry, and the expression of the isoforms in ovarian cancer cells with or without exposure to the tumor microenvironment, i.e., the ascitic fluid, was determined by western blotting assays. Immunohistochemical results suggest that PHF20L1 exhibits increased expression in sections of tumor tissues from patients with ovarian cancer and that higher PHF20L1 expression correlates with shorter progression-free survival and shorter overall survival. Furthermore, western blotting assays determined that protein isoforms are differentially regulated in SKOV-3 cells in response to stimulation with ascites from patients with epithelial ovarian cancer. The results suggest that PHF20L1 could play a relevant role in ovarian cancer given that higher PHF20L1 protein expression is associated with lower overall patient survival.
Sigma-Aldrich
Anti-PHF20L1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2