Merck
  • In vivo reconstitution finds multivalent RNA-RNA interactions as drivers of mesh-like condensates.
In vivo reconstitution finds multivalent RNA-RNA interactions as drivers of mesh-like condensates.
eLife (2021-03-03)
Weirui Ma, Gang Zheng, Wei Xie, Christine Mayr
ABSTRACT
Liquid-like condensates have been thought to be sphere-like. Recently, various condensates with filamentous morphology have been observed in cells. One such condensate is the TIS granule network that shares a large surface area with the rough endoplasmic reticulum and is important for membrane protein trafficking. It has been unclear how condensates with mesh-like shapes but dynamic protein components are formed. In vitro and in vivo reconstitution experiments revealed that the minimal components are a multivalent RNA-binding protein that concentrates RNAs that are able to form extensive intermolecular mRNA-mRNA interactions. mRNAs with large unstructured regions have a high propensity to form a pervasive intermolecular interaction network that acts as condensate skeleton. The underlying RNA matrix prevents full fusion of spherical liquid-like condensates, thus driving the formation of irregularly shaped membraneless organelles. The resulting large surface area may promote interactions at the condensate surface and at the interface with other organelles.
Sigma-Aldrich
Anti-HuR Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid