Merck
  • Highly Expressed CYBRD1 Associated with Glioma Recurrence Regulates the Immune Response of Glioma Cells to Interferon.
Highly Expressed CYBRD1 Associated with Glioma Recurrence Regulates the Immune Response of Glioma Cells to Interferon.
Evidence-based complementary and alternative medicine : eCAM (2021-07-31)
Mingjie Qing, Jiahao Zhou, Weijian Chen, Lijuan Cheng
ABSTRACT
Invasiveness, resistance to treatment, and recurrence of gliomas are significant hurdles to successful treatment regimens. Data sets from Gene Expression Omnibus (GEO), CGGA-RNAseq, and The Cancer Genome Atlas Glioblastoma Multiforme (TCGA-GBM) were analyzed, and an increased expression of Cytochrome B Reductase 1 (CYBRD1) was identified and could be associated with aggravated clinical outcomes. Gene ontology (GO) enrichment analysis indicated that CYBRD1 co-expressed genes are enriched during an immune response. CYBRD1 overexpression in glioma cell lines is enhanced, whereas CYBRD1 silencing attenuated the aggressiveness of glioma cells. In IFN-α-treated glioma cells, IFN-α suppressed the viability and migratory ability and invasive ability of glioma cells, whereas CYBRD1 overexpression attenuated the antitumor effects of IFN-α. CYBRD1 could potentially serve as a biomarker for glioma recurrence. CYBRD1 overexpression enhances glioma cell aggressiveness and attenuates glioma cell response to IFN-α.
Sigma-Aldrich
Anti-CYBRD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution