Merck
  • Downregulation of the Ca2+-activated K+ channel KCa3.1 in mouse preosteoblast cells treated with vitamin D receptor agonist.
Downregulation of the Ca2+-activated K+ channel KCa3.1 in mouse preosteoblast cells treated with vitamin D receptor agonist.
American journal of physiology. Cell physiology (2020-06-11)
Hiroaki Kito, Haruka Morihiro, Yuka Sakakibara, Kyoko Endo, Junko Kajikuri, Takayoshi Suzuki, Susumu Ohya
ABSTRACT
The maturity of osteoblasts by proliferation and differentiation in preosteoblasts is essential for maintaining bone homeostasis. The beneficial effects of vitamin D on bone homeostasis in mammals have been demonstrated experimentally and clinically. However, the direct actions of vitamin D on preosteoblasts remain to be fully elucidated. In this study, we found that the functional activity of intermediate-conductance Ca2+-activated K+ channels (KCa3.1) positively regulated cell proliferation in MC3T3-E1 cells derived from mouse preosteoblasts by enhancing intracellular Ca2+ signaling. We examined the effects of treatment with vitamin D receptor (VDR) agonist on the expression and activity of KCa3.1 by real-time PCR examination, Western blotting, Ca2+ imaging, and patch clamp analyses in mouse MC3T3-E1 cells. Following the downregulation of KCa3.1 transcriptional modulators such as Fra-1 and HDAC2, KCa3.1 activity was suppressed in MC3T3-E1 cells treated with VDR agonists. Furthermore, application of the KCa3.1 activator DCEBIO attenuated the VDR agonist-evoked suppression of cell proliferation rate. These findings suggest that a decrease in KCa3.1 activity is involved in the suppression of cell proliferation rate in VDR agonist-treated preosteoblasts. Therefore, KCa3.1 plays an important role in bone formation by promoting osteoblastic proliferation under physiological conditions.
Sigma-Aldrich
DCEBIO, ≥98% (HPLC), solid
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid