Merck
  • Hydrangea-like mesoporous WO3 nanoflowers with crystalline framework for 3-hydroxy-2-butanone sensing.
Hydrangea-like mesoporous WO3 nanoflowers with crystalline framework for 3-hydroxy-2-butanone sensing.
Analytical and bioanalytical chemistry (2020-10-04)
Dongpo Xu, Kangjie Ge, Shuyan Qi, Yan Chen, Jingxuan Qiu, Qing Liu
ABSTRACT
In this study, a simple and efficient strategy for the construction of hydrangea-like mesoporous WO3 nanoflowers templated using diblock copolymer PS119-PtBA129 was developed. The nanoflower shows good gas sensing properties, especially for 3-hydroxy-2-butanone (3H-2B), which is the signature metabolite of Listeria monocytogenes (L. monocytogenes). Therefore, the gas sensing of 3H-2B by hydrangea-like mesoporous WO3 nanoflowers can be used to detect L. monocytogenes. In the case of 25 ppm 3H-2B as target gas, the response (Ra/Rg) of the hydrangea-like mesoporous WO3 nanoflowers at 205 °C is 152, where Ra and Rg are the resistances of the sensing device in air and target gas, respectively, and the response and recovery times at 25 ppm are 25 s and 146 s, respectively. Schematic illustration of the formation of hydrangea-like mesoporous WO3 nanoflowers and its gas sensing implication.
Sigma-Aldrich
Ethyl α-bromoisobutyrate, 98%
Sigma-Aldrich
tert-Butyl acrylate, contains 10-20 ppm monomethyl ether hydroquinone as inhibitor, 98%