Merck
  • Epidemiology of systemic sclerosis: a multi-database population-based study in Tuscany (Italy).
Epidemiology of systemic sclerosis: a multi-database population-based study in Tuscany (Italy).
Orphanet journal of rare diseases (2021-02-19)
Alessio Coi, Simone Barsotti, Michele Santoro, Fabio Almerigogna, Elena Bargagli, Marzia Caproni, Giacomo Emmi, Bruno Frediani, Serena Guiducci, Marco Matucci Cerinic, Marta Mosca, Paola Parronchi, Renato Prediletto, Enrico Selvi, Gabriele Simonini, Antonio Gaetano Tavoni, Fabrizio Bianchi, Anna Pierini
ABSTRACT
Systemic Sclerosis (SSc) is a chronic autoimmune disease with a complex pathogenesis that includes vascular injury, abnormal immune activation, and tissue fibrosis. We provided a complete epidemiological characterization of SSc in the Tuscany region (Italy), considering prevalence and incidence, survival, comorbidities and drug prescriptions, by using a multi-database population-based approach. Cases of SSc diagnosed between 1st January 2003 and 31st December 2017 among residents in Tuscany were collected from the population-based Rare Diseases Registry of Tuscany. All cases were linked to regional health and demographic databases to obtain information about vital statistics, principal causes of hospitalization, complications and comorbidities, and drug prescriptions. The prevalence of SSc in Tuscany population resulted to be 22.2 per 100,000, with the highest prevalence observed for the cases aged ≥ 65 years (33.2 per 100,000, CI 95% 29.6-37.3). In females, SSc was predominant (86.7% on the total) with an overall sex ratio F/M of 6.5. Nevertheless, males presented a more severe disease, with a lower survival and significant differences in respiratory complications and metabolic comorbidities. Complications and comorbidities such as pulmonary involvement (HR = 1.66, CI 95% 1.17-2.35), congestive heart failure (HR = 2.76, CI 95% 1.80-4.25), subarachnoid and intracerebral haemorrhage (HR = 2.33, CI 95% 1.21-4.48) and malignant neoplasms (HR = 1.63, CI 95% 1.06-2.52), were significantly associated to a lower survival, also after adjustment for age, sex and other SSc-related complications. Disease-modifying antirheumatic drugs, endothelin receptor antagonists, and phosphodiesterase-5 inhibitors were the drugs with the more increasing prevalence of use in the 2008-2017 period. The multi-database approach is important in the investigation of rare diseases where it is often difficult to provide accurate epidemiological indicators. A population-based registry can be exploited in synergy with health databases, to provide evidence related to disease outcomes and therapies and to assess the burden of disease, relying on a large cohort of cases. Building an integrated archive of data from multiple databases linking a cohort of patients to their comorbidities, clinical outcomes and survival, is important both in terms of treatment and prevention.
Sigma-Aldrich
Treprostinil, ≥98% (HPLC)