Merck
  • Opposite estrogen effects of estrone and 2-hydroxyestrone on MCF-7 sensitivity to the cytotoxic action of cell growth, oxidative stress and inflammation activity.
Opposite estrogen effects of estrone and 2-hydroxyestrone on MCF-7 sensitivity to the cytotoxic action of cell growth, oxidative stress and inflammation activity.
Ecotoxicology and environmental safety (2020-12-16)
Shiying Xu, Jiadi Sun, Yinzhi Zhang, Jian Ji, Xiulan Sun
ABSTRACT
There are many kinds of estrogens, and endogenous estrogens produce a variety of estrogen metabolites with similar structure but with different physiological effects after metabolism in vivo. Studies have shown that estrone (E1) widely occurs in the environment and animal-derived food. Because of its estrogen effect, E1 can have adverse effects on the human body as an endocrine disruptor. In this study, we found that E1 and 2-hydroxyestrone (2-OH-E1), the hydroxylation metabolite of estrogen, have opposite proliferative effects on breast cancer cells (MCF-7) through cell proliferation experiments and comparison of their effects by molecular docking and detection of ROS, Ca2+, and cell pathway proteins. The effects of 2-methoxyestrone (2-MeO-E1) and 16α-hydroxyestrone (16α-OH-E1) on the biochemical and protein levels of MCF-7 were further studied to compare the effects of metabolic sites and modes on estrogen effects. Hydroxylation of E1 at the C2 site weakened the estrogen effect, down-regulated the expression of the mammalian target of rapamycin (mTOR) and protein kinase B (Akt) pathway proteins, inhibited the proliferation of cancer cells, and enhanced anti-oxidative stress and anti-inflammation. Methoxylation at the C2 position also inhibited the expression of inflammatory and oxidative stress pathway proteins but did not greatly affect the estrogen effects. However, hydroxylation on C16 had no significant effect on the biological effects of estrogen. Therefore, the structural changes of estrogen on C2 are important reasons for the different physiological effects of estrogen and its metabolites. Thus, by regulating the gene Cytochrome P450 1B1(CYP1B1), which affects the hydroxylation metabolism of estrogen, and promoting the hydroxylation of estrone at the C2 position, the estrogen effect of estrone can be effectively reduced, thus reducing the harm its poses in food and the environment.