Merck
  • Morphological and biochemical characterization of remodeling in aorta and vena cava of DOCA-salt hypertensive rats.
Morphological and biochemical characterization of remodeling in aorta and vena cava of DOCA-salt hypertensive rats.
American journal of physiology. Heart and circulatory physiology (2007-01-24)
Stephanie W Watts, Catherine Rondelli, Keshari Thakali, Xiaopeng Li, Bruce Uhal, Mohammad H Pervaiz, Ralph E Watson, Gregory D Fink
ABSTRACT
Arterial remodeling occurs in response to mechanical and neurohumoral stimuli. We hypothesized that veins, which are not exposed to higher pressures in hypertension, would demonstrate less active remodeling than arteries. We assessed remodeling with two standard measures of arterial remodeling: vessel morphometry and the expression/function of matrix metalloproteinases (MMPs). Thoracic aorta and vena cava from sham normotensive and DOCA-salt hypertensive rats (110 +/- 4 and 188 +/- 8 mmHg systolic blood pressure, respectively) were used. Wall thickness was increased in DOCA-salt vs. sham aorta (301 +/- 23 vs. 218 +/- 14 mum, P < 0.05), as was medial area, but neither measure was altered in the vena cava. The aorta and vena cava expressed the gelatinases MMP-2, MMP-9, transmembrane proteinase MT1-MMP, and tissue inhibitor of metalloproteinase-2 (TIMP-2). Immunohistochemically, MMP-2 localized to smooth muscle in the aorta and densely in endothelium/smooth muscle of the vena cava. Western and zymographic analyses verified that MMP-2 was active in all vessels and less active in the vena cava than aorta. In hypertension, MMP-2 expression and activity in the aorta were increased (59.1 +/- 3.7 and 74.5 +/- 6.1 units in sham and DOCA, respectively, P < 0.05); similar elevations were not observed in the vena cava. MMP-9 was weakly expressed in all vessels. MT1-MMP was expressed by the aorta and vena cava and elevated in the vena cava from DOCA-salt rats. TIMP-2 expression was significantly increased in the aorta of DOCA rats compared with sham but was barely detectable in the vena cava of sham or DOCA-salt hypertensive rats. These findings suggest that large veins may not undergo vascular remodeling in DOCA-salt hypertension.
Sigma-Aldrich
Anti-MT1-MMP (Ab-4) Mouse mAb (113-5B7), liquid, clone 113-5B7, Calbiochem®