Merck
  • A model combining age, equivalent uniform dose and IL-8 may predict radiation esophagitis in patients with non-small cell lung cancer.
A model combining age, equivalent uniform dose and IL-8 may predict radiation esophagitis in patients with non-small cell lung cancer.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology (2018-03-03)
Shulian Wang, Jeff Campbell, Matthew H Stenmark, Paul Stanton, Jing Zhao, Martha M Matuszak, Randall K Ten Haken, Feng-Ming Kong
ABSTRACT
To study whether cytokine markers may improve predictive accuracy of radiation esophagitis (RE) in non-small cell lung cancer (NSCLC) patients. A total of 129 patients with stage I-III NSCLC treated with radiotherapy (RT) from prospective studies were included. Thirty inflammatory cytokines were measured in platelet-poor plasma samples. Logistic regression was performed to evaluate the risk factors of RE. Stepwise Akaike information criterion (AIC) and likelihood ratio test were used to assess model predictions. Forty-nine of 129 patients (38.0%) developed grade ≥2 RE. Univariate analysis showed that age, stage, concurrent chemotherapy, and eight dosimetric parameters were significantly associated with grade ≥2 RE (p < 0.05). IL-4, IL-5, IL-8, IL-13, IL-15, IL-1α, TGFα and eotaxin were also associated with grade ≥2 RE (p < 0.1). Age, esophagus generalized equivalent uniform dose (EUD), and baseline IL-8 were independently associated grade ≥2 RE. The combination of these three factors had significantly higher predictive power than any single factor alone. Addition of IL-8 to toxicity model significantly improves RE predictive accuracy (p = 0.019). Combining baseline level of IL-8, age and esophagus EUD may predict RE more accurately. Refinement of this model with larger sample sizes and validation from multicenter database are warranted.