Merck
  • Inhibition of clathrin-mediated endocytosis by knockdown of AP-2 leads to alterations in the plasma membrane proteome.
Inhibition of clathrin-mediated endocytosis by knockdown of AP-2 leads to alterations in the plasma membrane proteome.
Traffic (Copenhagen, Denmark) (2020-11-24)
David Tobys, Lisa Maria Kowalski, Eva Cziudaj, Stefan Müller, Peter Zentis, Elke Pach, Paola Zigrino, Tobias Blaeske, Stefan Höning
ABSTRACT
In eukaryotic cells, clathrin-mediated endocytosis (CME) is a central pathway for the internalization of proteins from the cell surface, thereby contributing to the maintenance of the plasma membrane protein composition. A key component for the formation of endocytic clathrin-coated vesicles (CCVs) is AP-2, as it sequesters cargo membrane proteins, recruits a multitude of other endocytic factors and initiates clathrin polymerization. Here, we inhibited CME by depletion of AP-2 and explored the consequences for the plasma membrane proteome. Quantitative analysis revealed accumulation of major constituents of the endosomal-lysosomal system reflecting a block in retrieval by compensatory CME. The noticeable enrichment of integrins and blockage of their turnover resulted in severely impaired cell migration. Rare proteins such as the anti-cancer drug target CA9 and tumor markers (CD73, CD164, CD302) were significantly enriched. The AP-2 knockdown attenuated the global endocytic capacity, but clathrin-independent entry pathways were still operating, as indicated by persistent internalization of specific membrane-spanning and GPI-anchored receptors (PVR, IGF1R, CD55, TNAP). We hypothesize that blocking AP-2 function and thus inhibiting CME may be a novel approach to identify new druggable targets, or to increase their residence time at the plasma membrane, thereby increasing the probability for efficient therapeutic intervention.
Sigma-Aldrich
Anti-CD73 Antibody, clone AA60-E3-3, clone AA60-E3-3, from mouse
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse