- Serum Anti-Collagen IV IgM and IgG Antibodies as Indicators of Low Vascular Turnover of Collagen IV in Patients with Long-Term Complications of Type 2 Diabetes.
Serum Anti-Collagen IV IgM and IgG Antibodies as Indicators of Low Vascular Turnover of Collagen IV in Patients with Long-Term Complications of Type 2 Diabetes.
Thickening of the vascular basement membrane (BM) is a fundamental structural change in the small blood vessels in diabetes. Collagen type IV (CIV) is a major component of the BMs, and monitoring the turnover of this protein in type 2 diabetes (T2D) can provide important information about the mechanisms of vascular damage. The aim of the study was through the use of non-invasive biomarkers of CIV (autoantibodies, derivative peptides, and immune complexes) to investigate vascular turnover of CIV in patients with long-term complications of T2D. We measured serum levels of these biomarkers in 59 T2D patients with micro- and/or macrovascular complications and 20 healthy controls using an ELISA. Matrix metalloproteinases-2 and -9 (MMP-2 and MMP-9) were also tested. In the T2D group, significantly lower levels of CIV markers and significantly higher levels of MMP-2 and MMP-9 were found compared to controls. A significant positive correlation was found between IgM antibody levels against CIV and MMP-2. These findings suggest that vascular metabolism of CIV is decreased in T2D with long-term complications and show that a positive linear relationship exists between MMP-2 levels and CIV turnover in the vascular wall.