Merck
  • Behavioral validation of a wireless low-power neurostimulation technology in a conditioned place preference task.
Behavioral validation of a wireless low-power neurostimulation technology in a conditioned place preference task.
Journal of neural engineering (2019-01-09)
Lisa Y Maeng, Maria F Murillo, Michelle Mu, Meng-Chen Lo, Marjorie de la Rosa, Jonathan M O'Brien, Daniel K Freeman, Alik S Widge
ABSTRACT
Neurostimulation technologies are important for studying neural circuits and the connections that underlie neurological and psychiatric disorders. However, current methods come with limitations such as the restraint on movement imposed by the wires delivering stimulation. The objective of this study was to assess whether the e-Particle (EP), a novel wireless neurostimulator, could sufficiently stimulate the brain to modify behavior without these limitations. Rats were implanted with the EP and a commercially available stimulating electrode. Animals received rewarding brain stimulation, and performance in a conditioned place preference (CPP) task was measured. To ensure stimulation-induced neuronal activation, immediate early gene c-fos expression was also measured. The EP was validated in a commonly used CPP task by demonstrating that (1) wireless stimulation via the EP induced preference behavior that was comparable to that induced by standard wired electrodes and (2) neuronal activation was observed in projection targets of the stimulation site. The EP may help achieve a better understanding of existing brain stimulation methods while overcoming their limitations. Validation of the EP in a behavioral model suggests that the benefits of this technology may extend to other areas of animal research and potentially to human clinical applications.
Sigma-Aldrich
Anti-c-Fos (Ab-2) (4-17) Rabbit pAb, lyophilized, Calbiochem®