- Oligometastatic Prostate Adenocarcinoma. Clinical-Pathologic Study of a Histologically Under-Recognized Prostate Cancer.
Oligometastatic Prostate Adenocarcinoma. Clinical-Pathologic Study of a Histologically Under-Recognized Prostate Cancer.
The clinical parameters and the histological and immunohistochemical findings of a prospective protocolized series of 27 prostate carcinoma patients with oligometastatic disease followed homogeneously were analyzed. Lymph nodes (81.5%) and bones (18.5%) were the only metastatic sites. Local control after metastatic directed treatment was achieved in 22 (81.5%) patients. A total of 8 (29.6%) patients developed castration-resistant prostate cancer. Seventeen (63%) patients presented with non-organ confined disease. The Gleason index 8-10 was the most frequently observed (12 cases, 44.4%) combined grade. Positive immunostainings were detected with androgen receptor (100%), PGP 9.5 (74%), ERG (40.7%), chromogranin A (29.6%), and synaptophysin (18.5%) antibodies. The Ki-67 index value > 5% was observed in 15% of the cases. L1CAM immunostaining was negative in all cases. Fisher exact test showed that successful local control of metastases was associated to mild inflammation, organ confined disease, Ki-67 index < 5%, and Gleason index 3 + 3. A castration resistant status was associated with severe inflammation, atrophy, a Gleason index higher than 3 + 3, Ki-67 index ≥ 5%, and positive PGP 9.5, chromogranin A, and synaptophysin immunostainings. In conclusion, oligometastatic prostate adenocarcinoma does not have a specific clinical-pathologic profile. However, some histologic and immunohistochemical parameters of routine use may help with making therapeutic decisions.