Merck
  • Reduction of nuclear Y654-p-β-catenin expression through SH3GL2-meditated downregulation of EGFR in chemotolerance TNBC: Clinical and prognostic importance.
Reduction of nuclear Y654-p-β-catenin expression through SH3GL2-meditated downregulation of EGFR in chemotolerance TNBC: Clinical and prognostic importance.
Journal of cellular physiology (2020-01-22)
Md Saimul Islam, Hemantika Dasgupta, Mukta Basu, Anup Roy, Neyaz Alam, Susanta Roychoudhury, Chinmay Kumar Panda
ABSTRACT
Triple negative breast cancer (TNBC) originates from a less differentiated ductal cell of breast, which is less sensitive to chemotherapy. The chemotolerance mechanism of TNBC has not yet been studied in detail. For this reason, molecular profiles (expression/genetic/epigenetic) of Y654-p-β-catenin (active) and its kinase epidermal growth factor receptor (EGFR) along with SH3GL2 (regulator of EGFR homeostasis) were compared between neoadjuvant chemotherapy treated (NACT) and pretherapeutic TNBC samples. Reduced nuclear expression of Y654-p-β-catenin protein with low proliferation index and CD44 prevalence showed concordance with reduced expression of EGFR/Y1045-p-EGFR proteins in the NACT samples than the pretherapeutic TNBC samples. Infrequent messenger RNA expression, gene amplification (10-32.5%), and mutation (1%) of EGFR were seen in the TNBC samples irrespective of therapy, suggesting the importance of EGFR protein stabilization in this tumor. The upregulation of SH3GL2 seen in the NACT samples in contrast to the pretherapeutic samples might be due to its promoter hypomethylation, as seen in the quantitative methylation assay. A similar trend of upregulation of SH3GL2 and downregulation of EGFR, Y1045-p-EGFR, Y654-p-β-catenin were seen in the MDA-MB-231 cell line using antharacycline antitumor drugs (doxorubicin/nogalamycin). The NACT patients with reduced expression of Y654-p-β-catenin and/or EGFR and high expression of SH3GL2 showed comparatively better prognosis than the pretherapeutic patients. Thus, our study showed that reduced nuclear expression of Y654-p-β-catenin in NACT samples due to downregulation of EGFR protein through promoter hypomethylation-mediated upregulation of SH3GL2, resulting in low proliferation index/CD44 prevalence with better prognosis of the NACT patients, might have an important role in the chemotolerance of TNBC.