Merck
  • SARS-CoV-2 Cell Entry Factors ACE2 and TMPRSS2 Are Expressed in the Microvasculature and Ducts of Human Pancreas but Are Not Enriched in β Cells.
SARS-CoV-2 Cell Entry Factors ACE2 and TMPRSS2 Are Expressed in the Microvasculature and Ducts of Human Pancreas but Are Not Enriched in β Cells.
Cell metabolism (2020-11-19)
Katie C Coate, Jeeyeon Cha, Shristi Shrestha, Wenliang Wang, Luciana Mateus Gonçalves, Joana Almaça, Meghan E Kapp, Maria Fasolino, Ashleigh Morgan, Chunhua Dai, Diane C Saunders, Rita Bottino, Radhika Aramandla, Regina Jenkins, Roland Stein, Klaus H Kaestner, Golnaz Vahedi, Marcela Brissova, Alvin C Powers
ABSTRACT
Isolated reports of new-onset diabetes in individuals with COVID-19 have led to the hypothesis that SARS-CoV-2 is directly cytotoxic to pancreatic islet β cells. This would require binding and entry of SARS-CoV-2 into β cells via co-expression of its canonical cell entry factors, angiotensin-converting enzyme 2 (ACE2) and transmembrane serine protease 2 (TMPRSS2); however, their expression in human pancreas has not been clearly defined. We analyzed six transcriptional datasets of primary human islet cells and found that ACE2 and TMPRSS2 were not co-expressed in single β cells. In pancreatic sections, ACE2 and TMPRSS2 protein was not detected in β cells from donors with and without diabetes. Instead, ACE2 protein was expressed in islet and exocrine tissue microvasculature and in a subset of pancreatic ducts, whereas TMPRSS2 protein was restricted to ductal cells. These findings reduce the likelihood that SARS-CoV-2 directly infects β cells in vivo through ACE2 and TMPRSS2.
Product Description
Swagelok® Union Cross, Swagelok® 400-4, brass, 1/4 in. Swagelok
Sigma-Aldrich
Anti-TMPRSS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, laboratory grade