Merck
  • Evaluation of different teas against starch digestibility by mammalian glycosidases.
Evaluation of different teas against starch digestibility by mammalian glycosidases.
Journal of agricultural and food chemistry (2010-01-07)
Lee Wah Koh, Lin Ling Wong, Ying Yan Loo, Stefan Kasapis, Dejian Huang
ABSTRACT
Current work investigated the ability of different tea (green, oolong and black teas) in inhibiting human salivary alpha-amylase (HSA) and mammalian alpha-glucosidase (AGH). The inhibitory profiles were correlated to their major polyphenol content (theaflavins and catechins). The fully fermented black tea was demonstrated to be most potent in inhibiting HSA and AGH (IC50 of 0.42 to 0.67 and 0.56 to 0.58 mg of tea leaves/mL respectively). Its capability in retarding the digestion of a real food system (rice noodle) was further elucidated with an in vitro digestion study. Results indicated that black tea was able to retard starch digestion moderately, thereby allowing a gradual reduction of sugar liberation. Polyphenolic profile analysis suggested that the oxidized catechins, theaflavins, may be responsible for its activity. We have found that refractive index (RI) measurement is a rapid, direct, and highly convenient method for quantifying the degree of enzymatic starch digestion and kinetics. The RI method has good linearity range, limit of detection (0.1596 mg/mL, maltose equivalent) and limit of quantitation (0.6312 mg/mL) and was successfully applied in our study.
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea