Merck
  • Narciclasine exerts anti-inflammatory actions by blocking leukocyte-endothelial cell interactions and down-regulation of the endothelial TNF receptor 1.
Narciclasine exerts anti-inflammatory actions by blocking leukocyte-endothelial cell interactions and down-regulation of the endothelial TNF receptor 1.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2019-04-25)
Anna Stark, Rebecca Schwenk, Gesine Wack, Gabriele Zuchtriegel, Melissa G Hatemler, Jacqueline Bräutigam, Achim Schmidtko, Christoph A Reichel, Iris Bischoff, Robert Fürst
ABSTRACT
The alkaloid narciclasine has been characterized extensively as an anticancer compound. Accumulating evidence suggests that narciclasine has anti-inflammatory potential; however, the underlying mechanism remains poorly understood. We hypothesized that narciclasine affects the activation of endothelial cells (ECs), a hallmark of inflammatory processes, which is a prerequisite for leukocyte-EC interaction. Thus, we aimed to investigate narciclasine's action on this process in vivo and to analyze the underlying mechanisms in vitro. In a murine peritonitis model, narciclasine reduced leukocyte infiltration, proinflammatory cytokine expression, and inflammation-associated abdominal pain. Moreover, narciclasine decreased rolling and blocked adhesion and transmigration of leukocytes in vivo. In cultured ECs, narciclasine inhibited the expression of cell adhesion molecules intercellular adhesion molecule-1, vascular cell adhesion molecule-1, and E-selectin and blocked crucial steps of the NF-κB activation cascade: NF-κB promotor activity, p65 nuclear translocation, inhibitor of κB α (IκBα) phosphorylation and degradation, and IκBα kinase β and TGF-β-activated kinase 1 phosphorylation. Interestingly, these effects were based on the narciclasine-triggered loss of TNF receptor 1 (TNFR1). Our study highlights narciclasine as an interesting anti-inflammatory compound that effectively inhibits the interaction of leukocytes with ECs by blocking endothelial activation processes. Most importantly, we showed that the observed inhibitory action of narciclasine on TNF-triggered signaling pathways is based on the loss of TNFR1.-Stark, A., Schwenk, R., Wack, G., Zuchtriegel, G., Hatemler, M. G., Bräutigam, J., Schmidtko, A., Reichel, C. A., Bischoff, I., Fürst, R. Narciclasine exerts anti-inflammatory actions by blocking leukocyte-endothelial cell interactions and down-regulation of the endothelial TNF receptor 1.
Sigma-Aldrich
Narciclasine, ≥98% (HPLC)