Merck
  • Thermodynamic, Physical, and Structural Characteristics in Layered Hybrid Type (C2H5NH3)2MCl4 (M = 59Co, 63Cu, 65Zn, and 113Cd) Crystals.
Thermodynamic, Physical, and Structural Characteristics in Layered Hybrid Type (C2H5NH3)2MCl4 (M = 59Co, 63Cu, 65Zn, and 113Cd) Crystals.
Molecules (Basel, Switzerland) (2020-04-25)
Ae Ran Lim
ABSTRACT
The thermal, physical, and molecular dynamics of layered hybrid type (C2H5NH3)2MCl4 (M = 59Co, 63Cu, 65Zn, and 113Cd) crystals were investigated by thermogravimetric analysis (TGA) and magic angle spinning nuclear magnetic resonance (MAS NMR) spectroscopy. The temperatures of the onset of partial thermal decomposition were found to depend on the identity of M. In addition, the Bloembergen-Purcell-Pound curves for the 1H spin-lattice relaxation time T1ρ in the rotating frames of CH3CH2 and NH3, and for the 13C T1ρ of CH3 and CH2 were shown to exhibit minima as a function of the inverse temperature. These results confirmed the rotational motion of 1H and 13C in the C2H5NH3 cation. Finally, the T1ρ values and activation energies Ea obtained from the 1H measurements for the H‒Cl···M (M = Zn and Cd) bond in the absence of paramagnetic ions were larger than those obtained for the H‒Cl···M (M = Co and Cu) bond in the presence of paramagnetic ions. Moreover, the Ea value for 13C, which is distant from the M ions, was found to decrease upon increasing the mass of the M ion, unlike in the case of the Ea values for 1H.
Sigma-Aldrich
Allyl methyl sulfone, 96%