Merck
  • Chemical Evaluation of the Effects of Storage Conditions on the Botanical Goldenseal using Marker-based and Metabolomics Approaches.
Chemical Evaluation of the Effects of Storage Conditions on the Botanical Goldenseal using Marker-based and Metabolomics Approaches.
The Yale journal of biology and medicine (2020-07-02)
Manead Khin, Nadja B Cech, Joshua J Kellogg, Lindsay K Caesar
ABSTRACT
Hydrastis canadensis, commonly known as goldenseal, is a botanical native to the southeastern United States that has been used for the treatment of infection. The activity of goldenseal is often attributed to the presence of alkaloids (cyclic, nitrogen-containing compounds) present within its roots. Chemical components of botanical supplements like goldenseal may face degradation if not stored properly. The purpose of the research was to analyze the stability of known and unknown metabolites of H. canadensis during exposure to different storage conditions using mass spectrometry. Three abundant metabolites of H. canadensis, berberine, canadine, and hydrastine, were chosen for targeted analysis, and the stability of unknown metabolites was evaluated using untargeted metabolomics. The analysis and evaluation of H. canadensis samples were performed utilizing LC-MS and Principal Component Analysis (PCA). The research project focused on identifying the chemical changes in the metabolite content of H. canadensis under different temperature conditions (40°C ± 5°C, 20°C ± 5°C , and 4°C ± 5°C), different light:dark (hr:hr) cycles (16:8, 12:12, and 0:24), and different sample conditions (powdered roots versus whole roots) over a six month period. The results of this 6-month study revealed that the storage conditions evaluated had no significant effects on the chemical composition of H. canadensis roots. Hence, as long as H. canadensis roots are stored within the storage conditions tested in the study, no significant changes in chemical compositions of metabolites are expected.
Product Description
Canadine, phyproof® Reference Substance